Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MIS18A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018944.2 | NP_061817.1 | 2 | 431 | ||
XM_017028400.1 | XP_016883889.1 | 2 | 522 | |||
XM_017028401.1 | XP_016883890.1 | 2 | 522 | |||
XM_017028402.1 | XP_016883891.1 | 2 | 130 | |||
GenBank mRNA | AF231921.1 | - | 2 | 403 | ||
AF387845.1 | - | 2 | 401 | |||
AK075281.1 | - | 2 | 403 | |||
AK312073.1 | - | 2 | 522 | |||
BC042917.1 | - | 2 | 407 | 5934336 | ||
BU616573.1 | - | 2 | 434 | |||
CB959520.1 | - | 480 |