Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNTG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018968.3 | NP_061841.2 | 2 | 217 | ||
XM_011510366.2 | XP_011508668.1 | 2 | 369 | |||
XM_017004361.1 | XP_016859850.1 | 2 | 179 | |||
XM_017004362.1 | XP_016859851.1 | 2 | 185 | |||
XM_017004363.1 | XP_016859852.1 | 2 | 371 | |||
XM_017004365.1 | XP_016859854.1 | 2 | 180 | |||
XM_017004366.1 | XP_016859855.1 | 2 | 180 | |||
XM_017004367.1 | XP_016859856.1 | 2 | 217 | |||
XM_017004368.1 | XP_016859857.1 | 2 | 180 | |||
XM_017004369.1 | XP_016859858.1 | 2 | 180 | |||
XM_017004370.1 | XP_016859859.1 | 2 | 180 | |||
XM_017004371.1 | XP_016859860.1 | 2 | 181 | |||
XM_017004372.1 | XP_016859861.1 | 2 | 181 | |||
XM_017004374.1 | XP_016859863.1 | 2 | 186 | |||
GenBank mRNA | AJ003029.1 | - | 2 | 219 | ||
BC125251.1 | - | 2 | 146 | 40025232 |