Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GDAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040875.2 | NP_001035808.1 | 2 | 152 | ||
NM_018972.2 | NP_061845.2 | 2 | 282 | |||
XM_011517551.2 | XP_011515853.1 | 2 | 1414 | |||
XM_017013585.1 | XP_016869074.1 | 2 | 1414 | |||
XM_017013586.1 | XP_016869075.1 | 2 | 1414 | |||
GenBank mRNA | AB551556.1 | - | 2 | 281 | ||
AB551557.1 | - | 2 | 281 | |||
AB551558.1 | - | 2 | 279 | |||
AK292572.1 | - | 2 | 221 | |||
BC024939.1 | - | 2 | 117 | 3923335 | ||
BC035450.1 | - | 2 | 129 | |||
DB505741.1 | - | 2 | 282 | |||
Y17849.1 | - | 2 | 210 |