Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAJC10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271581.1 | NP_001258510.1 | 8 | 1320 | ||
NM_018981.2 | NP_061854.1 | 8 | 1320 | |||
NR_073365.1 | 8 | 1320 | ||||
NR_073366.1 | 8 | 1320 | ||||
NR_073367.1 | 8 | 1320 | ||||
GenBank mRNA | AF038503.1 | - | 8 | 1031 | ||
AF490904.1 | - | 8 | 1068 | |||
AK027450.1 | - | 6 | 661 | |||
AK027647.1 | - | 8 | 1089 | |||
AK027696.1 | - | 8 | 1068 | |||
AK027701.2 | - | 5 | 442 | |||
AK298138.1 | - | 4 | 305 | |||
AL832267.1 | - | 1 | 20 | |||
AL832632.2 | - | 8 | 1091 | |||
AL832646.3 | - | 6 | 672 | |||
AY089971.1 | - | 6 | 674 | |||
AY358577.1 | - | 8 | 1077 | |||
BC070034.1 | - | 8 | 1065 | |||
BC107425.1 | - | 6 | 697 | |||
BC117299.1 | - | 8 | 1035 | 40125850 | ||
BC126168.1 | - | 8 | 1035 | 8991884 |