Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TOLLIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318512.1 | NP_001305441.1 | 5 | 3375 | ||
NM_001318514.1 | NP_001305443.1 | 6 | 3535 | |||
NM_001318515.1 | NP_001305444.1 | 3 | 3098 | |||
NM_001318516.1 | NP_001305445.1 | 5 | 3342 | |||
NM_019009.3 | NP_061882.2 | 6 | 3525 | |||
XM_017017931.1 | XP_016873420.1 | 4 | 3432 | |||
GenBank mRNA | AB209541.1 | - | 7 | 3230 | ||
AK096693.1 | - | 5 | 3300 | |||
AK127123.1 | - | 6 | 3489 | |||
AL136835.1 | - | 6 | 3472 | |||
BX648136.1 | - | 1 | 3802 |