Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SGTB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019072.2 | NP_061945.1 | 11 | 1522 | ||
XM_005248548.3 | XP_005248605.1 | 11 | 1389 | |||
XM_017009597.1 | XP_016865086.1 | 12 | 1658 | |||
GenBank mRNA | AK096321.1 | - | 11 | 1477 |