Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DLL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019074.3 | NP_061947.1 | 10 | 2294 | ||
GenBank mRNA | AB036931.1 | - | 10 | 2234 | ||
AB043894.1 | - | 10 | 1958 | |||
AF253468.1 | - | 10 | 1958 | |||
AF279305.1 | - | 9 | 1864 | |||
AY358894.1 | - | 10 | 1993 | |||
BC106950.2 | - | 10 | 2094 | 40034887 |