Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPOP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007226.1 | NP_001007227.1 | 5 | 563 | ||
NM_001007227.1 | NP_001007228.1 | 4 | 441 | |||
NM_001007228.1 | NP_001007229.1 | 3 | 389 | |||
NM_001007229.1 | NP_001007230.1 | 4 | 440 | |||
NM_001007230.1 | NP_001007231.1 | 5 | 492 | |||
NM_003563.3 | NP_003554.1 | 4 | 511 | |||
XM_005257723.4 | XP_005257780.1 | 4 | 735 | |||
XM_005257724.4 | XP_005257781.1 | 3 | 665 | |||
XM_017025204.1 | XP_016880693.1 | 2 | 178 | |||
GenBank mRNA | AJ000644.1 | - | 3 | 249 | ||
AK222589.1 | - | 3 | 214 | |||
AK293045.1 | - | 4 | 722 | |||
AK293587.1 | - | 3 | 331 | |||
AK312691.1 | - | 3 | 225 | |||
BC001269.1 | - | 4 | 395 | 3461565 | ||
BC003385.1 | - | 3 | 206 | 3445878 | ||
CB992730.1 | - | 5 | 464 |