Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRLS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127458.1 | NP_001120930.1 | 6 | 600 | ||
NM_001323561.1 | NP_001310490.1 | 6 | 608 | |||
NM_001323562.1 | NP_001310491.1 | 6 | 615 | |||
NM_001323563.1 | NP_001310492.1 | 6 | 983 | |||
NM_001323564.1 | NP_001310493.1 | 6 | 609 | |||
NM_019095.5 | NP_061968.1 | 6 | 921 | |||
NR_136617.1 | 5 | 835 | ||||
XM_005260738.4 | XP_005260795.1 | 6 | 585 | |||
GenBank mRNA | AF241784.1 | - | 6 | 840 | ||
AL832419.1 | - | 6 | 668 | |||
BC069010.1 | - | 5 | 758 | 5092704 | ||
BM563835.1 | - | 6 | 603 | |||
BU852955.1 | - | 7 | 628 | |||
DQ386730.1 | - | 6 | 747 |