Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GTPBP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286216.1 | NP_001273145.1 | 7 | 844 | ||
NM_019096.4 | NP_061969.3 | 7 | 1039 | |||
XM_017010976.1 | XP_016866465.1 | 7 | 1033 | |||
GenBank mRNA | AB024574.1 | - | 8 | 882 | ||
AF168990.1 | - | 7 | 836 | |||
AJ420518.1 | - | 2 | 202 | |||
AK290267.1 | - | 7 | 844 | |||
AL834331.1 | - | 7 | 821 | |||
BC020980.2 | - | 3 | 455 | |||
BC028347.2 | - | 5 | 747 | |||
BC064968.1 | - | 7 | 992 | 6149902 |