Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GTPBP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286216.1 | NP_001273145.1 | 12 | 2577 | ||
NM_019096.4 | NP_061969.3 | 12 | 2772 | |||
GenBank mRNA | AB024574.1 | - | 13 | 2614 | ||
AF168990.1 | - | 12 | 2569 | |||
AJ420518.1 | - | 7 | 1935 | |||
AK000430.1 | - | 9 | 1387 | |||
AL834331.1 | - | 12 | 2554 | |||
BC020980.2 | - | 8 | 2188 | |||
BC028347.2 | - | 10 | 2480 | |||
BC064968.1 | - | 12 | 2725 | 6149902 |