Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPXM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001184699.1 | NP_001171628.1 | 4 | 547 | ||
NM_019609.4 | NP_062555.1 | 4 | 547 | |||
GenBank mRNA | AK027661.1 | - | 4 | 483 | ||
AK075527.1 | - | 4 | 522 | |||
AK303142.1 | - | 1 | 26 | |||
AK315027.1 | - | 4 | 534 | |||
AL541188.3 | - | 4 | 519 | |||
AY358956.1 | - | 4 | 503 | |||
BC063430.1 | - | 4 | 524 | 5239864 | ||
BX366559.2 | - | 4 | 529 | |||
CN308423.1 | - | 4 | 538 |