Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCL28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301873.1 | NP_001288802.1 | 2 | 178 | ||
NM_001301874.1 | NP_001288803.1 | 2 | 178 | |||
NM_001301875.1 | NP_001288804.1 | 2 | 269 | |||
NM_148672.3 | NP_683513.1 | 2 | 178 | |||
GenBank mRNA | AF110384.1 | - | 2 | 157 | ||
AF220210.1 | - | 2 | 103 | |||
AF266504.1 | - | 2 | 155 | |||
AK307307.1 | - | 2 | 133 | |||
AK312120.1 | - | 2 | 161 | |||
BC062668.1 | - | 2 | 120 | 30326117 | ||
BC069532.1 | - | 2 | 153 | 7262110 | ||
BX486632.1 | - | 2 | 179 | |||
CD671712.1 | - | 2 | 269 | |||
HM067700.1 | - | 2 | 103 |