Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FSTL5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128427.2 | NP_001121899.1 | 10 | 1643 | ||
NM_001128428.2 | NP_001121900.1 | 10 | 1643 | |||
NM_020116.4 | NP_064501.2 | 10 | 1646 | |||
XM_011532126.1 | XP_011530428.1 | 10 | 1612 | |||
GenBank mRNA | AB033089.1 | - | 10 | 1573 | ||
AK315152.1 | - | 10 | 1614 | |||
AL137695.1 | - | 7 | 989 | |||
BC036502.2 | - | 10 | 1647 | 5265170 |