Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZC3HAV1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020119.3 | NP_064504.2 | 4 | 1136 | ||
NM_024625.3 | NP_078901.3 | 4 | 1136 | |||
XM_005250501.2 | XP_005250558.1 | 4 | 1137 | |||
XM_011516412.2 | XP_011514714.1 | 4 | 1137 | |||
GenBank mRNA | AK055851.1 | - | 4 | 771 | ||
AK292811.1 | - | 4 | 804 | |||
BC025308.1 | - | 4 | 1065 | 5418915 | ||
BC033105.1 | - | 4 | 1065 | 5418956 | ||
BC040956.1 | - | 4 | 1136 | 5531412 | ||
BX571742.1 | - | 4 | 1135 | |||
BX647974.1 | - | 4 | 772 |