Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZC3HAV1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020119.3 | NP_064504.2 | 5 | 1941 | ||
NM_024625.3 | NP_078901.3 | 5 | 1941 | |||
XM_005250501.2 | XP_005250558.1 | 5 | 2308 | |||
XM_011516412.2 | XP_011514714.1 | 5 | 1942 | |||
GenBank mRNA | AF138863.1 | - | 2 | 246 | ||
AK023350.1 | - | 2 | 314 | |||
AK055851.1 | - | 4 | 2392 | |||
AK292811.1 | - | 5 | 1609 | |||
BC002935.1 | - | 2 | 654 | |||
BC025308.1 | - | 5 | 1870 | 5418915 | ||
BC033105.1 | - | 5 | 1870 | 5418956 | ||
BC040956.1 | - | 5 | 1941 | 5531412 | ||
BX571742.1 | - | 5 | 1940 | |||
BX647138.1 | - | 2 | 672 | |||
BX647974.1 | - | 5 | 1577 |