Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UGGT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020120.3 | NP_064505.1 | 3, 4 | 462 | ||
NR_027671.2 | 3, 4 | 615 | ||||
XM_006712634.3 | XP_006712697.1 | 3, 4 | 436 | |||
XM_006712635.3 | XP_006712698.1 | 3, 4 | 430 | |||
XM_006712636.3 | XP_006712699.1 | 3, 4 | 425 | |||
XM_017004507.1 | XP_016859996.1 | 4, 5 | 521 | |||
XM_017004508.1 | XP_016859997.1 | 4, 5 | 476 | |||
GenBank mRNA | AF227905.1 | - | 3, 4 | 322 | ||
AK293066.1 | - | 3, 4 | 615 | |||
BC041098.1 | - | 3, 4 | 612 | 5492334 | ||
BP356698.1 | - | 3, 4 | 461 | |||
DB226703.1 | - | 3, 4 | 459 |