Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UGGT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020120.3 | NP_064505.1 | 5, 6 | 709 | ||
NR_027671.2 | 5, 6 | 862 | ||||
XM_006712634.3 | XP_006712697.1 | 5, 6 | 683 | |||
XM_006712635.3 | XP_006712698.1 | 5, 6 | 677 | |||
XM_006712636.3 | XP_006712699.1 | 5, 6 | 672 | |||
XM_017004507.1 | XP_016859996.1 | 6, 7 | 768 | |||
XM_017004508.1 | XP_016859997.1 | 6, 7 | 723 | |||
GenBank mRNA | AF227905.1 | - | 5, 6 | 569 | ||
AK293066.1 | - | 5, 6 | 862 | |||
AK309101.1 | - | 3, 4 | 434 | |||
BC041098.1 | - | 5, 6 | 859 | 5492334 |