Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLAMF8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020125.2 | NP_064510.1 | 5 | 1064 | ||
XM_011509777.2 | XP_011508079.1 | 5 | 2177 | |||
XM_011509780.2 | XP_011508082.2 | 4 | 671 | |||
XM_017001844.1 | XP_016857333.1 | 6 | 2093 | |||
XM_017001845.1 | XP_016857334.1 | 5 | 1127 | |||
XM_017001846.1 | XP_016857335.1 | 5 | 951 | |||
GenBank mRNA | AF144235.1 | - | 2 | 145 | ||
AF146761.1 | - | 5 | 942 | |||
AK074669.1 | - | 5 | 1003 | |||
BC109194.2 | - | 5 | 880 | 40004601 |