Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SH3GLB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287045.1 | NP_001273974.1 | 12 | 1252 | ||
NM_001287046.1 | NP_001273975.1 | 10 | 1225 | |||
NM_020145.3 | NP_064530.1 | 10 | 1225 | |||
XM_005252098.2 | XP_005252155.1 | 11 | 1208 | |||
XM_005252100.2 | XP_005252157.1 | 10 | 1144 | |||
XM_005252101.3 | XP_005252158.1 | 8 | 883 | |||
XM_006717188.2 | XP_006717251.1 | 12 | 1296 | |||
XM_006717189.2 | XP_006717252.1 | 11 | 1279 | |||
XM_006717190.1 | XP_006717253.1 | 11 | 1281 | |||
XM_006717191.1 | XP_006717254.1 | 11 | 1209 | |||
GenBank mRNA | AB058751.2 | - | 12 | 1178 | ||
AF257319.1 | - | 10 | 1086 | |||
AF258589.1 | - | 10 | 1139 | |||
AF370426.1 | - | 3 | 1761 | |||
AF370433.1 | - | 3 | 997 | |||
AL832592.1 | - | 3 | 4115 | |||
BC014635.1 | - | 10 | 1172 | 3677306 |