Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SH3GLB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287045.1 | NP_001273974.1 | 10 | 981 | ||
NM_001287046.1 | NP_001273975.1 | 9 | 969 | |||
NM_020145.3 | NP_064530.1 | 9 | 969 | |||
XM_005252098.2 | XP_005252155.1 | 9 | 937 | |||
XM_005252100.2 | XP_005252157.1 | 8 | 873 | |||
XM_005252101.3 | XP_005252158.1 | 7 | 627 | |||
XM_006717188.2 | XP_006717251.1 | 10 | 1025 | |||
XM_006717189.2 | XP_006717252.1 | 9 | 1008 | |||
XM_006717190.1 | XP_006717253.1 | 10 | 1025 | |||
XM_006717191.1 | XP_006717254.1 | 10 | 953 | |||
GenBank mRNA | AB058751.2 | - | 10 | 907 | ||
AF257319.1 | - | 9 | 830 | |||
AF258589.1 | - | 9 | 884 | |||
AF370426.1 | - | 2 | 1505 | |||
AF370433.1 | - | 2 | 741 | |||
AL832592.1 | - | 2 | 3859 | |||
BC014635.1 | - | 9 | 916 | 3677306 |