Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STARD7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020151.3 | NP_064536.2 | 8 | 2917 | ||
XM_011511499.1 | XP_011509801.1 | 8 | 2534 | |||
GenBank mRNA | AB075479.1 | - | 1 | 544 | ||
AF270647.1 | - | 8 | 2844 | |||
AK023234.1 | - | 7 | 2177 | |||
AK054615.1 | - | 8 | 2283 | |||
AK094925.1 | - | 2 | 2489 | |||
AK096666.1 | - | 5 | 1677 | |||
BC003028.1 | - | 8 | 2898 | |||
BC007894.2 | - | 9 | 2675 | 4134933 | ||
BC008894.2 | - | 8 | 2898 | 3833465 | ||
BC009998.2 | - | 9 | 2675 | 4128869 | ||
BC012774.2 | - | 8 | 2864 | 3842611 | ||
BC012793.2 | - | 8 | 2878 | 4080200 | ||
BC013279.2 | - | 8 | 2898 | 3830320 | ||
BC014076.2 | - | 8 | 2878 | 4127007 | ||
BC014274.2 | - | 8 | 2896 | 3677198 | ||
BC032106.1 | - | 8 | 2890 | 4643387 | ||
CR749695.1 | - | 1 | 1232 |