Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OTOR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020157.3 | NP_064542.1 | 3, 4 | 416 | ||
GenBank mRNA | AF233261.1 | - | 3, 4 | 411 | ||
AF243505.1 | - | 3, 4 | 374 | |||
AJ242552.1 | - | 3, 4 | 367 | |||
AY359082.1 | - | 3, 4 | 404 | |||
BC101688.1 | - | 3, 4 | 416 | 8069194 | ||
BC101690.1 | - | 3, 4 | 416 | 8069196 |