Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMARCAD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128429.2 | NP_001121901.1 | 10 | 1653 | ||
NM_001128430.1 | NP_001121902.1 | 10 | 1555 | |||
NM_001254949.1 | NP_001241878.1 | 2 | 475 | |||
NM_020159.4 | NP_064544.2 | 10 | 1653 | |||
NR_045644.1 | 11 | 1704 | ||||
XM_017008463.1 | XP_016863952.1 | 10 | 1477 | |||
XM_017008464.1 | XP_016863953.1 | 10 | 1477 | |||
XM_017008465.1 | XP_016863954.1 | 10 | 1487 | |||
GenBank mRNA | AB032948.2 | - | 9 | 1441 | ||
AK025302.1 | - | 5 | 693 | |||
AK027490.1 | - | 10 | 1551 | |||
AK301668.1 | - | 2 | 475 | |||
AL359929.1 | - | 1 | 83 | |||
AY008271.1 | - | 10 | 1456 | |||
BC045534.1 | - | 10 | 1608 | 5267669 |