Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAD18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020165.3 | NP_064550.3 | 12 | 1464 | ||
XM_017006873.1 | XP_016862362.1 | 13 | 1622 | |||
GenBank mRNA | AB035274.2 | - | 12 | 1499 | ||
AF169796.1 | - | 12 | 1444 | |||
AK023075.1 | - | 12 | 1451 | |||
AK222771.1 | - | 12 | 1442 | |||
AY004333.1 | - | 12 | 1410 | |||
BC001302.1 | - | 12 | 1431 | 3451960 |