Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC177 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271507.1 | NP_001258436.1 | 2 | 3992 | ||
GenBank mRNA | AF284217.1 | - | 1 | 1201 |