Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PMEPA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001255976.1 | NP_001242905.1 | 4 | 1212 | ||
NM_020182.4 | NP_064567.2 | 4 | 1483 | |||
NM_199169.2 | NP_954638.1 | 4 | 1136 | |||
NM_199170.2 | NP_954639.1 | 4 | 1129 | |||
NM_199171.2 | NP_954640.1 | 4 | 1194 | |||
GenBank mRNA | AF224278.1 | - | 4 | 1080 | ||
AF305616.1 | - | 4 | 1410 | |||
AK296199.1 | - | 4 | 1135 | |||
AY128643.1 | - | 4 | 1084 |