Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OLFML3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286352.1 | NP_001273281.1 | 4 | 1788 | ||
NM_001286353.1 | NP_001273282.1 | 3 | 1765 | |||
NM_020190.3 | NP_064575.1 | 3 | 1636 | |||
XM_017001848.1 | XP_016857337.1 | 3 | 1798 | |||
GenBank mRNA | AF201945.1 | - | 3 | 1570 | ||
AK075353.1 | - | 3 | 1538 | |||
AK075369.1 | - | 3 | 1584 | |||
AK075479.1 | - | 3 | 1549 | |||
AK075544.1 | - | 2 | 1867 | |||
AK092766.1 | - | 4 | 1738 | |||
AK222471.1 | - | 3 | 1553 | |||
AK223221.1 | - | 3 | 1566 | |||
AY358954.1 | - | 3 | 1619 | |||
AY464015.1 | - | 3 | 1601 | |||
BC009920.2 | - | 3 | 1561 | 3010136 |