Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C14orf132 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252507.2 | NP_001239436.1 | 2 | 7335 | ||
NM_001282463.1 | NP_001269392.1 | 3 | 7434 | |||
NM_001282464.1 | NP_001269393.1 | 3 | 576 | |||
NM_001289139.1 | NP_001276068.1 | 3 | 7428 | |||
XM_017021469.1 | XP_016876958.1 | 2 | 8667 | |||
GenBank mRNA | AF131832.1 | - | 1 | 1525 | ||
AK022021.1 | - | 1 | 2267 | |||
AL390130.1 | - | 1 | 3213 | |||
BC042922.1 | - | 1 | 3212 | |||
BC043593.1 | - | 2 | 7315 | |||
BI791809.1 | - | 3 | 487 | |||
DB264577.1 | - | 1 | 142 |