Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRDM10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020228.2 | NP_064613.2 | 6 | 780 | ||
NM_199437.1 | NP_955469.1 | 6 | 780 | |||
NM_199438.1 | NP_955470.1 | 2 | 506 | |||
NM_199439.1 | NP_955471.1 | 2 | 506 | |||
XM_011542912.2 | XP_011541214.1 | 6 | 1042 | |||
XM_017018023.1 | XP_016873512.1 | 2 | 2591 | |||
GenBank mRNA | AB033057.2 | - | 2 | 290 | ||
AF275817.1 | - | 2 | 506 | |||
AF503171.1 | - | 2 | 437 | |||
AK310354.1 | - | 7 | 826 | |||
AL833172.1 | - | 6 | 783 | |||
BC112934.1 | - | 6 | 772 | 6502888 | ||
BC117415.1 | - | 2 | 345 | 40125966 | ||
BC143612.1 | - | 2 | 345 | 9052125 | ||
BC143613.1 | - | 2 | 345 | 9052126 | ||
BX648944.1 | - | 6 | 781 |