Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BBX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142568.2 | NP_001136040.1 | 11 | 1989 | ||
NM_020235.6 | NP_064620.2 | 11 | 1989 | |||
XM_005247642.3 | XP_005247699.1 | 11 | 1905 | |||
XM_005247643.3 | XP_005247700.1 | 11 | 1917 | |||
XM_005247644.3 | XP_005247701.1 | 12 | 2080 | |||
XM_011512999.2 | XP_011511301.1 | 11 | 1989 | |||
XM_011513000.1 | XP_011511302.1 | 11 | 2117 | |||
XM_011513001.1 | XP_011511303.1 | 11 | 1957 | |||
XM_011513002.2 | XP_011511304.1 | 11 | 1989 | |||
XM_011513003.2 | XP_011511305.1 | 11 | 1989 | |||
XM_011513004.2 | XP_011511306.1 | 11 | 2117 | |||
XM_011513005.2 | XP_011511307.1 | 11 | 1989 | |||
XM_017006881.1 | XP_016862370.1 | 13 | 2130 | |||
XM_017006882.1 | XP_016862371.1 | 12 | 2080 | |||
GenBank mRNA | AF276948.1 | - | 8 | 1687 | ||
AF454941.1 | - | 11 | 1982 | |||
AF454942.1 | - | 9 | 1862 | |||
AK025944.1 | - | 1 | 240 | |||
AK026082.1 | - | 1 | 321 | |||
AK130627.1 | - | 1 | 302 | |||
AK291757.1 | - | 11 | 1935 | |||
AK314434.1 | - | 11 | 1940 | |||
AL136769.1 | - | 9 | 1862 | |||
AL834307.1 | - | 1 | 299 | |||
BC012837.1 | - | 1 | 631 |