Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC4A7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF047033.1 | - | 3 | 288 | ||
AF089726.1 | - | 3 | 288 | |||
EU934248.1 | - | 3 | 465 |