Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAK5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020341.3 | NP_065074.1 | 5 | 951 | ||
NM_177990.2 | NP_817127.1 | 4 | 845 | |||
XM_017027960.1 | XP_016883449.1 | 6 | 1060 | |||
XM_017027961.1 | XP_016883450.1 | 7 | 1115 | |||
XM_017027962.1 | XP_016883451.1 | 6 | 1006 | |||
XM_017027963.1 | XP_016883452.1 | 4 | 1019 | |||
XM_017027964.1 | XP_016883453.1 | 4 | 1401 | |||
XM_017027965.1 | XP_016883454.1 | 4 | 708 | |||
GenBank mRNA | AB033090.1 | - | 5 | 953 | ||
AB040812.1 | - | 5 | 786 | |||
AK095674.1 | - | 5 | 743 | |||
AK291401.1 | - | 4 | 676 | |||
AK307926.1 | - | 4 | 776 | |||
AK315820.1 | - | 4 | 846 | |||
AX776013.1 | - | 5 | 953 | |||
BC024179.1 | - | 4 | 684 | 4821164 |