Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AVEN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020371.2 | NP_065104.1 | 5 | 958 | ||
XM_011521818.2 | XP_011520120.1 | 6 | 1141 | |||
XM_011521819.2 | XP_011520121.1 | 5 | 1184 | |||
XM_011521820.1 | XP_011520122.1 | 5 | 659 | |||
GenBank mRNA | AF283508.1 | - | 5 | 954 | ||
BC010488.1 | - | 5 | 952 | 2900586 | ||
BC063533.1 | - | 5 | 942 | 4425400 |