Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RGS5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195303.2 | NP_001182232.1 | 2 | 437 | ||
NM_001254748.1 | NP_001241677.1 | 5 | 495 | |||
NM_001254749.1 | NP_001241678.1 | 4 | 610 | |||
NM_003617.3 | NP_003608.1 | 4 | 610 | |||
GenBank mRNA | AB008109.1 | - | 4 | 353 | ||
AF030108.1 | - | 4 | 348 | |||
AF159570.1 | - | 4 | 316 | |||
AF493929.1 | - | 4 | 272 | |||
AJ891044.1 | - | 2 | 99 | |||
AK315357.1 | - | 4 | 353 | |||
AU130764.1 | - | 4 | 519 | |||
BC030059.1 | - | 4 | 335 | 4098248 | ||
BP292867.1 | - | 5 | 455 | |||
BU172751.1 | - | 4 | 340 | |||
BX537427.1 | - | 4 | 354 |