Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: REXO4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001279350.1 | NP_001266279.1 | 4 | 889 | ||
NM_001279351.1 | NP_001266280.1 | 4 | 750 | |||
NM_020385.3 | NP_065118.2 | 4 | 1067 | |||
NR_103996.1 | 3 | 720 | ||||
GenBank mRNA | AF273304.1 | - | 4 | 1015 | ||
AF295774.1 | - | 4 | 1015 | |||
AK025493.1 | - | 4 | 921 | |||
AK295982.1 | - | 4 | 750 | |||
AK304547.1 | - | 4 | 805 | |||
AK314333.1 | - | 4 | 981 | |||
AK315904.1 | - | 5 | 740 | |||
AL136894.1 | - | 4 | 998 | |||
BC009274.2 | - | 4 | 970 | 3958331 | ||
BC071691.1 | - | 3 | 579 |