Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HRASLS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020386.4 | NP_065119.2 | 4 | 829 | ||
XM_017006923.1 | XP_016862412.1 | 5 | 710 | |||
GenBank mRNA | AB030816.1 | - | 4 | 634 | ||
AB510981.1 | - | 3 | 423 | |||
AY251533.1 | - | 3 | 436 | |||
BC048095.1 | - | 4 | 832 | 4830228 |