Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPL47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020409.2 | NP_065142.2 | 3 | 294 | ||
NM_177988.1 | NP_817125.1 | 2 | 148 | |||
GenBank mRNA | AY212270.1 | - | 3 | 271 | ||
BC021575.1 | - | 2 | 148 | 3161966 | ||
BC032522.1 | - | 3 | 227 | 5500168 | ||
CA428787.1 | - | 3 | 307 |