Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DDX24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020414.3 | NP_065147.1 | 4 | 1412 | ||
GenBank mRNA | AB209382.1 | - | 3 | 1332 | ||
AF134475.1 | - | 2 | 228 | |||
AF145022.1 | - | 2 | 228 | |||
AF214731.1 | - | 4 | 1411 | |||
AK025162.1 | - | 4 | 1408 | |||
AK296726.1 | - | 5 | 1123 | |||
AK297232.1 | - | 4 | 1276 | |||
AK304504.1 | - | 3 | 679 | |||
AL136886.1 | - | 4 | 1388 | |||
BC008847.2 | - | 4 | 1388 | 4301025 | ||
BC009406.2 | - | 4 | 1388 | 4303448 | ||
BC096826.1 | - | 5 | 1218 | 30925011 | ||
BX537533.1 | - | 4 | 1399 |