Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PCBP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001174100.1 | NP_001167571.1 | 8 | 802 | ||
NM_033008.2 | NP_127501.1 | 7 | 654 | |||
NM_033010.2 | NP_127503.1 | 7 | 596 | |||
XM_005265330.2 | XP_005265387.1 | 8 | 2609 | |||
XM_005265331.2 | XP_005265388.1 | 8 | 728 | |||
XM_006713268.2 | XP_006713331.1 | 8 | 2417 | |||
XM_006713269.1 | XP_006713332.1 | 9 | 873 | |||
XM_006713271.2 | XP_006713334.1 | 8 | 2609 | |||
XM_017006916.1 | XP_016862405.1 | 9 | 1030 | |||
XM_017006917.1 | XP_016862406.1 | 8 | 2417 | |||
XM_017006918.1 | XP_016862407.1 | 7 | 2269 | |||
XM_017006919.1 | XP_016862408.1 | 7 | 645 | |||
GenBank mRNA | AF092441.1 | - | 7 | 568 | ||
AF176330.1 | - | 7 | 596 | |||
AF257770.1 | - | 4 | 1233 | |||
AF257771.1 | - | 5 | 1068 | |||
AK023993.1 | - | 7 | 643 | |||
AK225815.1 | - | 8 | 780 | |||
BC003008.1 | - | 7 | 617 | 2822683 | ||
BC004153.2 | - | 7 | 617 | 2822683 | ||
BX647811.1 | - | 6 | 1183 |