Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PELI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020651.3 | NP_065702.2 | 7 | 1287 | ||
XM_005264437.3 | XP_005264494.1 | 6 | 1106 | |||
XM_011532994.2 | XP_011531296.1 | 7 | 1946 | |||
XM_011532996.2 | XP_011531298.1 | 7 | 2705 | |||
XM_011532997.2 | XP_011531299.1 | 7 | 1322 | |||
XM_017004520.1 | XP_016860009.1 | 8 | 1421 | |||
XM_017004521.1 | XP_016860010.1 | 8 | 1280 | |||
GenBank mRNA | AF300987.1 | - | 6 | 842 | ||
AF302505.1 | - | 6 | 4865 | |||
AJ278859.1 | - | 6 | 827 | |||
AK027668.1 | - | 4 | 598 | |||
AK312441.1 | - | 7 | 1236 | |||
AY364257.1 | - | 6 | 827 | |||
BC050019.1 | - | 7 | 1241 | 5296990 | ||
BC050533.1 | - | 7 | 1223 | |||
BC063611.1 | - | 7 | 1215 | 4992160 | ||
BX649152.1 | - | 3 | 2143 |