Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLK4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020666.2 | NP_065717.1 | 3 | 323 | ||
GenBank mRNA | AF294429.1 | - | 2 | 215 | ||
AK294921.1 | - | 3 | 322 | |||
AK304614.1 | - | 3 | 325 | |||
AK310427.1 | - | 4 | 1132 | |||
AK311485.1 | - | 3 | 325 | |||
BC015942.1 | - | 3 | 323 | |||
BC034633.1 | - | 3 | 277 | |||
BC136261.1 | - | 3 | 271 | 9020248 | ||
BC143547.1 | - | 3 | 271 | 9052058 | ||
BC151233.1 | - | 4 | 417 | |||
CR749504.1 | - | 5 | 1202 |