Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BCR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004327.3 | NP_004318.3 | 1 | 1174 | ||
NM_021574.2 | NP_067585.2 | 1 | 1174 | |||
GenBank mRNA | AB209991.1 | - | 1 | 923 | ||
BC066122.1 | - | 1 | 1005 | |||
EU216058.1 | - | 1 | 578 | |||
EU216059.1 | - | 1 | 578 | |||
EU216061.1 | - | 1 | 578 | |||
EU216062.1 | - | 1 | 578 | |||
EU216063.1 | - | 1 | 578 | |||
EU216064.1 | - | 1 | 578 | |||
EU216067.1 | - | 1 | 578 | |||
EU216069.1 | - | 1 | 578 | |||
EU216070.1 | - | 1 | 578 | |||
X02596.1 | - | 1 | 1066 | |||
X06418.1 | - | 621 | ||||
Y00661.1 | - | 1 | 1174 |