Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IGSF9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135050.1 | NP_001128522.1 | 21 | 3890 | ||
NM_020789.3 | NP_065840.2 | 21 | 3842 | |||
GenBank mRNA | AB037776.1 | - | 21 | 3890 | ||
AK056650.1 | - | 10 | 4051 | |||
AY203932.1 | - | 9 | 3408 | |||
AY358128.1 | - | 14 | 2522 | |||
BC030141.2 | - | 21 | 3842 | 4762143 |