Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARHGAP21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020824.3 | NP_065875.3 | 26 | 5207 | ||
XM_005252542.3 | XP_005252599.1 | 26 | 5275 | |||
XM_005252544.4 | XP_005252601.1 | 25 | 6930 | |||
XM_011519602.2 | XP_011517904.1 | 27 | 7062 | |||
XM_011519603.2 | XP_011517905.1 | 27 | 5469 | |||
XM_011519604.2 | XP_011517906.1 | 27 | 5285 | |||
XM_011519605.2 | XP_011517907.1 | 26 | 7032 | |||
XM_011519606.2 | XP_011517908.1 | 24 | 6927 | |||
XM_011519607.2 | XP_011517909.1 | 26 | 6956 | |||
XM_017016459.1 | XP_016871948.1 | 26 | 5367 | |||
XM_017016460.1 | XP_016871949.1 | 25 | 5337 | |||
XM_017016461.1 | XP_016871950.1 | 25 | 5148 | |||
GenBank mRNA | AB037845.2 | - | 25 | 4680 | ||
AF480466.1 | - | 26 | 5154 | |||
AL834495.1 | - | 18 | 2811 | |||
BC022931.1 | - | 1 | 155 | |||
BC118915.2 | - | 17 | 3158 | 40071194 | ||
BX537570.1 | - | 17 | 4905 |