Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SYT13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001247987.1 | NP_001234916.1 | 8 | 4321 | ||
NM_020826.2 | NP_065877.1 | 6 | 4103 | |||
GenBank mRNA | AB037848.1 | - | 6 | 4090 | ||
AL512743.1 | - | 6 | 4027 |