Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLHL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020866.2 | NP_065917.1 | 2 | 1311 | ||
XM_017020678.1 | XP_016876167.1 | 2 | 231 | |||
GenBank mRNA | AB040923.1 | - | 2 | 1277 | ||
AF252283.1 | - | 2 | 1145 | |||
AK054609.1 | - | 2 | 523 | |||
AK291435.1 | - | 2 | 1311 | |||
BC022460.1 | - | 2 | 555 | 4819077 |