Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SH3RF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020870.3 | NP_065921.2 | 9 | 1883 | ||
GenBank mRNA | AB040927.1 | - | 5 | 798 | ||
AK021429.1 | - | 9 | 1744 | |||
AK025255.1 | - | 5 | 715 | |||
BC033203.1 | - | 8 | 1430 | |||
BC041023.1 | - | 7 | 1066 | |||
BC053671.1 | - | 7 | 2225 | 6144754 |