Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001324188.1 | NP_001311117.1 | 3 | 3688 | ||
NM_001324189.1 | NP_001311118.1 | 3 | 3611 | |||
NM_020873.6 | NP_065924.3 | 2 | 3406 | |||
GenBank mRNA | AB040930.1 | - | 2 | 5905 | ||
AB074161.1 | - | 1 | 2639 | |||
AK023273.1 | - | 1 | 1793 | |||
BC034947.2 | - | 2 | 2969 | 4826337 |