Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USP28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301029.1 | NP_001287958.1 | 6 | 682 | ||
NM_020886.3 | NP_065937.1 | 8 | 893 | |||
XM_005271630.2 | XP_005271687.1 | 8 | 889 | |||
XM_005271631.2 | XP_005271688.1 | 8 | 889 | |||
XM_005271632.2 | XP_005271689.1 | 9 | 1219 | |||
XM_005271633.4 | XP_005271690.1 | 9 | 947 | |||
XM_005271636.2 | XP_005271693.1 | 8 | 889 | |||
XM_005271637.2 | XP_005271694.1 | 8 | 889 | |||
XM_005271638.2 | XP_005271695.1 | 8 | 889 | |||
XM_005271639.2 | XP_005271696.1 | 8 | 889 | |||
XM_011542936.2 | XP_011541238.1 | 7 | 808 | |||
XM_011542938.1 | XP_011541240.1 | 6 | 682 | |||
XM_017018056.1 | XP_016873545.1 | 7 | 808 | |||
XM_017018057.1 | XP_016873546.1 | 9 | 1219 | |||
XM_017018058.1 | XP_016873547.1 | 7 | 825 | |||
XM_017018059.1 | XP_016873548.1 | 7 | 808 | |||
XM_017018060.1 | XP_016873549.1 | 6 | 682 | |||
XM_017018061.1 | XP_016873550.1 | 6 | 682 | |||
XM_017018062.1 | XP_016873551.1 | 6 | 682 | |||
XM_017018065.1 | XP_016873554.1 | 8 | 857 | |||
XM_017018066.1 | XP_016873555.1 | 9 | 1219 | |||
GenBank mRNA | AF266283.1 | - | 8 | 830 | ||
AK097623.1 | - | 10 | 1006 | |||
AK304770.1 | - | 6 | 653 | |||
BC065928.1 | - | 8 | 851 | 6140725 |